Learn More
Cayman Chemical 4chloro2 5DMAhydrochlorI 10mg
SDP
Catalog No. 502954945
Encompass_Preferred
An analytical reference standard categorized as an amphetamine; has a high addictive liability in rodents; has been associated with fatality; has been detected in blotter papers seized by law enforcement; intended for research and forensic applications
Catalog No. 50-295-4945 Supplier Cayman Chemical Supplier No. 1203810 MG
An analytical reference standard categorized as an amphetamine; has a high addictive liability in rodents; has been associated with fatality; has been detected in blotter papers seized by law enforcement; intended for research and forensic applications