Learn More
Cayman Chemical 4-chloro-α-Cyanocinnamic Acid, Purity: ≥98%, CAS: 20374-46-3, 10 mg
SDP
Catalog No. 502893460
Encompass_Preferred
A phenylpropanoid which readily co-crystallizes with various biomolecules
Catalog No. 50-289-3460 Supplier Cayman Chemical Supplier No. 900098110 MG
A phenylpropanoid which readily co-crystallizes with various biomolecules