Learn More
Cayman Chemical 4-chloro-α-Pyrrolidinoisohexanophenone (hydrochloride), Purity: ≥98%, 5 mg
SDP
Catalog No. 502944755
Encompass_Preferred
An analytical reference standard categorized as a cathinone; intended for research and forensic applications
Catalog No. 50-294-4755 Supplier Cayman Chemical Supplier No. 410975 MG
An analytical reference standard categorized as a cathinone; intended for research and forensic applications