Learn More
Cayman Chemical 4chLRo Trazodon Isomerhy 10mg
SDP
Catalog No. 502913804
Encompass_Preferred
An isomer of trazodone; inhibits apomorphine-induced stereotypy in mice (ED50 = 0.24 mg/kg); induces catalepsy in mice (ED50 = 0.9 mg/kg)
Catalog No. 50-291-3804 Supplier Cayman Chemical Supplier No. 3540410 MG
An isomer of trazodone; inhibits apomorphine-induced stereotypy in mice (ED50 = 0.24 mg/kg); induces catalepsy in mice (ED50 = 0.9 mg/kg)