Learn More
Cayman Chemical 4-Chloromethamphetamine (hydrochloride), Purity: ≥98%, CAS: 30572-91-9, 50 mg
SDP
Catalog No. 502893656
Encompass_Preferred
A halogenated analog of methamphetamine; converted to 4-chloroamphetamine in vivo
Catalog No. 50-289-3656 Supplier Cayman Chemical Supplier No. 900218550 MG
A halogenated analog of methamphetamine; converted to 4-chloroamphetamine in vivo