Learn More
Cayman Chemical 4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline, Purity: ≥95%, CAS: 183322-18-1, 250 mg
SDP
Catalog No. 502990822
Encompass_Preferred
A building block and synthetic intermediate; has been used as a precursor in the synthesis of inhibitors, dual RTK and HDAC inhibitors, and anticancer compounds; a synthetic intermediate in the synthesis of EGFR inhibitors, including erlotinib, with antiproliferative activity
Catalog No. 50-299-0822 Supplier Cayman Chemical Supplier No. 30487250 MG
A building block and synthetic intermediate; has been used as a precursor in the synthesis of inhibitors, dual RTK and HDAC inhibitors, and anticancer compounds; a synthetic intermediate in the synthesis of EGFR inhibitors, including erlotinib, with antiproliferative activity