Learn More
Cayman Chemical 4CLOroIndole3ACtIc AcId 500mg
SDP
Catalog No. 502913617
Encompass_Preferred
An auxin plant growth regulator; induces tobacco callus growth and biologically active alkaloid production in Duboisia root cultures at 0.1 mg/ml; reduces heat stress-induced grain loss in wheat at 1 µM
Catalog No. 50-291-3617 Supplier Cayman Chemical Supplier No. 35534500 MG
An auxin plant growth regulator; induces tobacco callus growth and biologically active alkaloid production in Duboisia root cultures at 0.1 mg/ml; reduces heat stress-induced grain loss in wheat at 1 µM