Learn More
Cayman Chemical 4-Deoxypyridoxine Phosphate, Purity: ≥90%, CAS: 883-84-1, 10 mg
SDP
Catalog No. 502988772
Encompass_Preferred
A vitamin B6 analog and an inhibitor of S1P lyase; induces accumulation of [3H]dhS1P in IT-79MTNC3 cells that endogenously express high levels of S1P lyase (EC50s = 100 µM in the absence and presence of vitamin B6, respectively)
Catalog No. 50-298-8772 Supplier Cayman Chemical Supplier No. 2997310 MG
A vitamin B6 analog and an inhibitor of S1P lyase; induces accumulation of [3H]dhS1P in IT-79MTNC3 cells that endogenously express high levels of S1P lyase (EC50s = 100 µM in the absence and presence of vitamin B6, respectively)