Learn More
MSC GEARWRENCH Utility Tool Cart, 4-Drawer, Steel, 650 lb. Capacity, 1 ea.
Catalog No. 5000515281
Encompass
The 4-drawer utility tool cart is a heavy-duty storage solution engineered for professionals, featuring a robust steel construction with a powder-coated finish. This mobile cart offers a total load capacity of up to 650 lb and is designed to provide efficient organization and access to tools in demanding environments.
  • Four-drawer design featuring ball bearing slides rated for 100 lb loads
  • Trigger latch retention system for secure drawer closure
  • Five-inch by two-inch polyurethane casters with dual-locking top pedal brakes
  • External power tool rack and dedicated internal storage for long items
  • Full-length molded leg bumpers to minimize damage
  • Includes black EVA foam drawer liners for added tool protection
  • Integrated gas shocks for smooth opening
Catalog No. 50-005-15281 Supplier MSC Supplier No. 98154768
The 4-drawer utility tool cart is a heavy-duty storage solution engineered for professionals, featuring a robust steel construction with a powder-coated finish. This mobile cart offers a total load capacity of up to 650 lb and is designed to provide efficient organization and access to tools in demanding environments.
  • Four-drawer design featuring ball bearing slides rated for 100 lb loads
  • Trigger latch retention system for secure drawer closure
  • Five-inch by two-inch polyurethane casters with dual-locking top pedal brakes
  • External power tool rack and dedicated internal storage for long items
  • Full-length molded leg bumpers to minimize damage
  • Includes black EVA foam drawer liners for added tool protection
  • Integrated gas shocks for smooth opening