Learn More
Medchemexpress LLC (4E)-1,7-bis(4-hydroxy-3-methoxyphenyl)hept-4-en-3-one | 128700-97-0 | >95.0% | 1 ML
SDP
Catalog No. 5000230829
Encompass_Preferred
Gingerenone A is a diarylheptanoid research compound used as an Nrf2-Gpx4 activator and probe in studies of ferroptosis and inflammatory pathways. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available in solid quantities for analytical and research applications.
  • Natural diarylheptanoid with Nrf2-Gpx4 activity.
  • Supplied as a 10 mM solution in DMSO, 1 mL, for immediate use.
  • Also available in solid quantities for custom preparation.
  • Molecular formula C21H24O5 and molecular weight 356.41.
  • Intended for research use only; not for diagnostic or therapeutic use.
Catalog No. 50-002-30829 Supplier Medchemexpress LLC Supplier No. HY12091210MM 1ML
Gingerenone A is a diarylheptanoid research compound used as an Nrf2-Gpx4 activator and probe in studies of ferroptosis and inflammatory pathways. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available in solid quantities for analytical and research applications.
  • Natural diarylheptanoid with Nrf2-Gpx4 activity.
  • Supplied as a 10 mM solution in DMSO, 1 mL, for immediate use.
  • Also available in solid quantities for custom preparation.
  • Molecular formula C21H24O5 and molecular weight 356.41.
  • Intended for research use only; not for diagnostic or therapeutic use.