Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (4E)-7-(4-hydroxyphenyl)-1-phenylhept-4-en-3-one | 100667-52-5 | 97.0% | 280.36 g/mol | C19H20O2 | 5 MG
SDP

Catalog No. 5000221827
Encompass_Preferred

(4E)-7-(4-hydroxyphenyl)-1-phenylhept-4-en-3-one is a diarylheptanoid small-molecule for laboratory research and analytical applications. The compound is supplied as a solid with molecular formula C19H20O2, molecular weight 280.36 g/mol, and an analytical purity of 97.0% (NMR). Store protected from light at low temperatures; consult the certificate of analysis for batch-specific handling and stability information.

  • Solid, white to off-white appearance.
  • Molecular formula C19H20O2.
  • Molecular weight 280.36 g/mol.
  • Analytical purity 97.0% (NMR).
  • Storage: keep container tightly sealed, protect from light; recommended -20°C for solid, in solvent -80°C for 6 months or -20°C for 1 month.
  • Intended for research and analytical use only; not for human therapeutic use.

Catalog No. 50-002-21827 Supplier Medchemexpress LLC Supplier No. HY1346505MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(4E)-7-(4-hydroxyphenyl)-1-phenylhept-4-en-3-one is a diarylheptanoid small-molecule for laboratory research and analytical applications. The compound is supplied as a solid with molecular formula C19H20O2, molecular weight 280.36 g/mol, and an analytical purity of 97.0% (NMR). Store protected from light at low temperatures; consult the certificate of analysis for batch-specific handling and stability information.

  • Solid, white to off-white appearance.
  • Molecular formula C19H20O2.
  • Molecular weight 280.36 g/mol.
  • Analytical purity 97.0% (NMR).
  • Storage: keep container tightly sealed, protect from light; recommended -20°C for solid, in solvent -80°C for 6 months or -20°C for 1 month.
  • Intended for research and analytical use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.