Learn More
Medchemexpress LLC 4E1RCat | 328998-25-0 | 99.0% | C28H18N2O6 | 100 MG
SDP
Catalog No. 5000354100
Encompass_Preferred
4E1RCat is an inhibitor of cap-dependent translation, inhibiting the eIF4E:eIF4GI interaction with an IC50 of approximately 4 μM. It interferes with eIF4G and 4E-BP binding, blocking capped mRNA translation activated by CDK1/CYCB1. This compound is for research use only and affects chemosensitivity and targets translation in mice models.
  • Inhibits cap-dependent translation
  • Interferes with eIF4E:eIF4GI interaction
  • Blocks capped mRNA translation
  • Affects chemosensitivity in specific tumor types in mice
  • Sensitizes lymphomas to doxorubicin
  • Targets translation in mice
Catalog No. 50-003-54100 Supplier Medchemexpress LLC Supplier No. HY14427100MG
4E1RCat is an inhibitor of cap-dependent translation, inhibiting the eIF4E:eIF4GI interaction with an IC50 of approximately 4 μM. It interferes with eIF4G and 4E-BP binding, blocking capped mRNA translation activated by CDK1/CYCB1. This compound is for research use only and affects chemosensitivity and targets translation in mice models.
  • Inhibits cap-dependent translation
  • Interferes with eIF4E:eIF4GI interaction
  • Blocks capped mRNA translation
  • Affects chemosensitivity in specific tumor types in mice
  • Sensitizes lymphomas to doxorubicin
  • Targets translation in mice