Learn More
Selleck Chemical LLC 4E1RCat-5mg
SDP
Catalog No. 502389768
Encompass_Preferred
4E1RCat is a dual inhibitor of eIF4E:eIF4G and eIF4E:4E-BP1 interaction, and inhibits the binding of eIF4G to eIF4E.
Catalog No. 50-238-9768 Supplier Selleck Chemical LLC Supplier No. S73705MG
4E1RCat is a dual inhibitor of eIF4E:eIF4G and eIF4E:4E-BP1 interaction, and inhibits the binding of eIF4G to eIF4E.