Learn More
Medchemexpress LLC HY-19831 100mg , 4EGI-1 CAS:315706-13-9 Purity:98%
SDP
Catalog No. 501493045
Encompass_Preferred
Medchemexpress, HY-19831 100mg 4EGI-1 CAS:315706-13-9 Formula:C18H12Cl2N4O4S Kd: 25 μM (eIF4E/eIF4G) Purity:98% 4EGI-1 is an inhibitor of eIF4E/eIF4G interaction, with a K d of 25 μM against eIF4E binding. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3045 Supplier Medchemexpress LLC Supplier No. HY19831100MG
Medchemexpress, HY-19831 100mg 4EGI-1 CAS:315706-13-9 Formula:C18H12Cl2N4O4S Kd: 25 μM (eIF4E/eIF4G) Purity:98% 4EGI-1 is an inhibitor of eIF4E/eIF4G interaction, with a K d of 25 μM against eIF4E binding. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.