Learn More
Medchemexpress LLC HY-19831 10mg , 4EGI-1 CAS:315706-13-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1983110MG
Encompass_Preferred
HY-19831 10mg 4EGI-1 CAS: 315706-13-9 4EGI-1 is an inhibitor of eIF4E/eIF4G interaction, with a Kd of 25 μM against eIF4E binding. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0239