Learn More
Enzo Life Sciences 4EGI-1 (2mg). CAS: 315706-13-9
SDP
Supplier:  Enzo Life Sciences ALX430155M002
Encompass_Preferred
Selective and cell permeable inhibitor of the interaction between the human eukaryotic translation initiation factors 4E (eIF4E) and 4G (eIF4G). Alternative name: alpha-[2-[4-(3,4-Dichlorophenyl)-2-thiazolyl]hydrazinylidene]-2-nitro-benzenepropanoic acid. Purity: ≥95%. Appearance: Brown solid. Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-9242