Learn More
Medchemexpress LLC 4F 4PP oxalate | 144734-36-1 | 98.9% | 429.48 g/mol | C24H28FNO5 | 1 ML
SDP
Catalog No. 5000211515
Encompass_Preferred
4F 4PP (oxalate) is the oxalate salt of a selective serotonin 5-HT2A receptor antagonist supplied for research use. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL), with a reported CAS number 144734-36-1 and high purity suitable for in vitro pharmacology and receptor binding studies.
  • Selective 5-HT2A antagonist with reported Ki ≈ 5.3 nM
  • Low affinity at 5-HT2C (Ki ≈ 620 nM)
  • Supplied as a 10 mM solution in DMSO, 1 mL
  • High purity suitable for research (≈98.9%)
  • Molecular formula C24H28FNO5; molecular weight 429.48 g/mol
Catalog No. 50-002-11515 Supplier Medchemexpress LLC Supplier No. HY10097010MM 1ML
4F 4PP (oxalate) is the oxalate salt of a selective serotonin 5-HT2A receptor antagonist supplied for research use. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL), with a reported CAS number 144734-36-1 and high purity suitable for in vitro pharmacology and receptor binding studies.
  • Selective 5-HT2A antagonist with reported Ki ≈ 5.3 nM
  • Low affinity at 5-HT2C (Ki ≈ 620 nM)
  • Supplied as a 10 mM solution in DMSO, 1 mL
  • High purity suitable for research (≈98.9%)
  • Molecular formula C24H28FNO5; molecular weight 429.48 g/mol