Learn More
Medchemexpress LLC 4F 4PP oxalate | 144734-36-1 | 98.9% | 429.48 g/mol | C24H28FNO5 | 50 MG
SDP
Catalog No. 5000211201
Encompass_Preferred
4F 4PP oxalate is the oxalate salt of 4F 4PP, a selective serotonin 5-HT2A receptor antagonist used in pharmacology and neuroscience research. It shows high affinity at 5-HT2A and substantially lower affinity at 5-HT2C, and is supplied as a high-purity research chemical for binding and functional assays, and pharmacological characterization.
  • High affinity for 5-HT2A receptors (Ki ≈ 5.3 nM).
  • Lower affinity for 5-HT2C receptors (Ki ≈ 620 nM).
  • High purity suitable for research use (≈98.9%).
  • Available in multiple solid sizes and DMSO solution formats.
  • Molecular weight 429.48 g/mol; CAS 144734-36-1.
Catalog No. 50-002-11201 Supplier Medchemexpress LLC Supplier No. HY10097050MG
4F 4PP oxalate is the oxalate salt of 4F 4PP, a selective serotonin 5-HT2A receptor antagonist used in pharmacology and neuroscience research. It shows high affinity at 5-HT2A and substantially lower affinity at 5-HT2C, and is supplied as a high-purity research chemical for binding and functional assays, and pharmacological characterization.
  • High affinity for 5-HT2A receptors (Ki ≈ 5.3 nM).
  • Lower affinity for 5-HT2C receptors (Ki ≈ 620 nM).
  • High purity suitable for research use (≈98.9%).
  • Available in multiple solid sizes and DMSO solution formats.
  • Molecular weight 429.48 g/mol; CAS 144734-36-1.