Learn More
Medchemexpress LLC 4F 4PP oxalate | 144734-36-1 | 98.9% | 429.49 g/mol | C24H28FNO5 | 100 MG
SDP
Catalog No. 5000211650
Encompass_Preferred
4F 4PP oxalate is a research reagent that acts as a selective antagonist of the serotonin 5-HT2A receptor. It displays high affinity for 5-HT2A (Ki ≈ 5.3 nM) and substantially lower affinity for 5-HT2C (Ki ≈ 620 nM). Supplied as the oxalate salt, it is intended for in vitro and preclinical research use only.
  • High affinity for 5-HT2A receptor (Ki ≈ 5.3 nM).
  • Low affinity for 5-HT2C receptor (Ki ≈ 620 nM).
  • Supplied as an oxalate salt for improved handling and stability.
  • High purity suitable for research (≈98.9%).
  • Available in small laboratory quantities (e.g., 5 mg-100 mg).
  • Intended for in vitro and preclinical research use only.
Catalog No. 50-002-11650 Supplier Medchemexpress LLC Supplier No. HY100970100MG
4F 4PP oxalate is a research reagent that acts as a selective antagonist of the serotonin 5-HT2A receptor. It displays high affinity for 5-HT2A (Ki ≈ 5.3 nM) and substantially lower affinity for 5-HT2C (Ki ≈ 620 nM). Supplied as the oxalate salt, it is intended for in vitro and preclinical research use only.
  • High affinity for 5-HT2A receptor (Ki ≈ 5.3 nM).
  • Low affinity for 5-HT2C receptor (Ki ≈ 620 nM).
  • Supplied as an oxalate salt for improved handling and stability.
  • High purity suitable for research (≈98.9%).
  • Available in small laboratory quantities (e.g., 5 mg-100 mg).
  • Intended for in vitro and preclinical research use only.