Learn More
Cayman Chemical 4F 4PPoxalate 10mg
SDP
Supplier:  Cayman Chemical 3500510 MG
Encompass_Preferred
A 5-HT2A receptor antagonist (Ki = 5.3 nM); selective for 5-HT2A receptors over 5-HT1C receptors (Ki = 620 nM); inhibits 5-HT-induced current in Xenopus oocytes expressing human 5-HT2A receptors (IC50 = 5.15 µM); spinal superfusion reduces the area of electrically stimulated spinal field potentials in a rat model of SNL-induced neuropathic pain at 100 µM
Catalog No. 50-291-0516