Learn More
Cayman Chemical 4-fluoro-α-Pyrrolidinopentiophenone (hydrochloride), Purity: ≥98%, CAS: 850352-31-7, 10 mg
SDP
Catalog No. 502972796
Encompass_Preferred
An isomer of α-PVP characterized by the addition of a fluorine group at the 4 position of the phenyl ring; intended for forensic and research applications
Catalog No. 50-297-2796 Supplier Cayman Chemical Supplier No. 1516610 MG
An isomer of α-PVP characterized by the addition of a fluorine group at the 4 position of the phenyl ring; intended for forensic and research applications