Learn More
Cayman Chemical 4'-fluoro-α-Pyrrolidinopropiophenone (hydrochloride), Purity: ≥95%, CAS: 2748590-18-1, 50 mg
SDP
Catalog No. 502972165
Encompass_Preferred
A derivative of α-PPP, characterized by the addition of a fluorine group at the 4 position of the phenyl ring; intended for forensic and research applications
Catalog No. 50-297-2165 Supplier Cayman Chemical Supplier No. 1444850 MG
A derivative of α-PPP, characterized by the addition of a fluorine group at the 4 position of the phenyl ring; intended for forensic and research applications