Learn More
Cayman Chemical 4FluoroethcathInonhydroch 50mg
SDP
Catalog No. 502951348
Encompass_Preferred
A structural analog of 4-FMC, with an ethyl group replacing the amino terminal methyl group; intended to be used for research and forensic applications
Catalog No. 50-295-1348 Supplier Cayman Chemical Supplier No. 1123150 MG
A structural analog of 4-FMC, with an ethyl group replacing the amino terminal methyl group; intended to be used for research and forensic applications