Learn More
Cayman Chemical 4-Fluoromethamphetamine (hydrochloride), Purity: ≥98%, CAS: 52063-62-4, 50 mg
SDP
Catalog No. 502894170
Encompass_Preferred
A stimulant drug related to methamphetamine and 4-fluoroamphetamine that has been identified as a component of designer drugs sold as “legal high” replacements for restricted substances; intended for use as a standard for the forensic analysis of samples that may contain this compound
Catalog No. 50-289-4170 Supplier Cayman Chemical Supplier No. 900107050 MG
A stimulant drug related to methamphetamine and 4-fluoroamphetamine that has been identified as a component of designer drugs sold as “legal high” replacements for restricted substances; intended for use as a standard for the forensic analysis of samples that may contain this compound