Learn More
Cayman Chemical 4FluoromethcathInon metabol
SDP
Catalog No. 502892417
Encompass_Preferred
A metabolite of flephedrone that features conversion of the β-keto group to β-hydroxy and is an enantiomeric mixture of the R,R and S,S orientations at carbons one and two, as in pseudoephedrine
Catalog No. 50-289-2417 Supplier Cayman Chemical Supplier No. 9001433500 UG
A metabolite of flephedrone that features conversion of the β-keto group to β-hydroxy and is an enantiomeric mixture of the R,R and S,S orientations at carbons one and two, as in pseudoephedrine