Learn More
Chemglass Life Sciences Triple Insulated Hose, 1-1/2" Beaded Pipe to 3/4" Male NPT, 4ft Length
SDP
Catalog No. 50872385
Encompass_Preferred
Triple Insulated Hose, 1-1/2" Beaded Pipe to 3/4" Male NPT, 4ft Length. Triple Insulated Flexible Hose. Flexible stainless steel tubing, insulated with fiberglass and silicone. Temperature Range -80 to 200¿C. One end has beaded pipe fitting and the other is a 3/4¿ Male NPT.Hoses Are Designed For Use With CG-1969-M High Flow Manifold Systems
Catalog No. 50-872-385 Supplier Chemglass Life Sciences Supplier No. CG1969B154
Triple Insulated Hose, 1-1/2" Beaded Pipe to 3/4" Male NPT, 4ft Length. Triple Insulated Flexible Hose. Flexible stainless steel tubing, insulated with fiberglass and silicone. Temperature Range -80 to 200¿C. One end has beaded pipe fitting and the other is a 3/4¿ Male NPT.Hoses Are Designed For Use With CG-1969-M High Flow Manifold Systems