Learn More
AirSea USA 4GV Boxes for non-specific inners, 4GV/X4/S, Int Dims: 7 7/8"" x 7 7/8"" x 13 6/8""
Catalog No. 501534831
Encompass_Preferred
4GV Boxes - ship up to 4kg/8.82lbs. of dangerous goods without vermiculite. 4GV fiberboard box suitable for shipping dangerous goods by road, sea or air inside various inner packaging. UN Mark: 4GV/X4/S. External Dimensions: 210mm x 210mm x 365mm. Sold complete with plastic liner bag and suitable for non-specific inner packagings. Biodegradable chips must be used as a dangerous goods packaging material when shipping items within this 4GV box range.
Catalog No. 50-153-4831 Supplier AirSea USA Supplier No. 1014
4GV Boxes - ship up to 4kg/8.82lbs. of dangerous goods without vermiculite. 4GV fiberboard box suitable for shipping dangerous goods by road, sea or air inside various inner packaging. UN Mark: 4GV/X4/S. External Dimensions: 210mm x 210mm x 365mm. Sold complete with plastic liner bag and suitable for non-specific inner packagings. Biodegradable chips must be used as a dangerous goods packaging material when shipping items within this 4GV box range.