Learn More
AirSea USA UN 4GV boxes for non-specific inners, 4GV/X65/S, Int Dims: 23 5/8"" x 23 5/8"" x 24 5/8""
Catalog No. 501534834
Encompass_Preferred
4GV Box for non-specific inners, UN/4GV/X65/S,Hazardous Chemical shipments up to 65kg/143 lbs
Catalog No. 50-153-4834 Supplier AirSea USA Supplier No. 842
4GV Box for non-specific inners, UN/4GV/X65/S,Hazardous Chemical shipments up to 65kg/143 lbs