Learn More
AirSea USA 4GV boxes for non-specific inners, 4GV/X7/S. Int Dims: 10 13/16"" x 10 13/16"" x 11 7/16""
Catalog No. 501534832
Encompass_Preferred
4GV Boxes - ship up to 7kg/15.43lbs. of dangerous goods without vermiculite. 4GV fiberboad box suitable for shipping dangerous goods by road, sea or air inside various inner packagings. UN Mark: 4GV/X7/S. External Dimensions: 285mm x 285mm x 305mm. Sold complete with plastic liner bag and suitable for non-specific inner packagings. Biodegradable chips must be used as a dangerous goods packaging material when shipping items within this 4GV box range.
Catalog No. 50-153-4832 Supplier AirSea USA Supplier No. 1015
4GV Boxes - ship up to 7kg/15.43lbs. of dangerous goods without vermiculite. 4GV fiberboad box suitable for shipping dangerous goods by road, sea or air inside various inner packagings. UN Mark: 4GV/X7/S. External Dimensions: 285mm x 285mm x 305mm. Sold complete with plastic liner bag and suitable for non-specific inner packagings. Biodegradable chips must be used as a dangerous goods packaging material when shipping items within this 4GV box range.