Learn More
AirSea USA 4GV Boxes for Non-specific inners, 4GV/X7/S, external Dimensions: 11.22" x 11.82" x 12.01" certified for Lithium Batteries, Glass, Metal and Plastic inner containers
Catalog No. NC2101127
Encompass_Preferred
4GV Fibreboard box suitable for shipping dangerous goods by road, sea or air inside various inner packagings. UN Mark: 4GV/X5/S. Sold complete with plastic liner bag and suitable for non-specific inner packagings. Vermiculite must be used as a dangerous goods packaging material when shipping items within this 4GV box range.
This item is not returnable. View return policy
Catalog No. NC2101127 Supplier AirSea USA Supplier No. 839
4GV Fibreboard box suitable for shipping dangerous goods by road, sea or air inside various inner packagings. UN Mark: 4GV/X5/S. Sold complete with plastic liner bag and suitable for non-specific inner packagings. Vermiculite must be used as a dangerous goods packaging material when shipping items within this 4GV box range.