Learn More
Medchemexpress LLC 4H-1-Benzopyran-4-one, 2-(2,4-dihydroxyphenyl)-5,7-dihydroxy-3-(3-methyl-2-buten-1-yl)- | 73343-42-7 | 354.35 | 1 MG
SDP
Catalog No. 5000416580
Encompass_Preferred
Albanin A, a flavonoid, suppresses glutamate release by decreasing Ca2+/calmodulin/adenylate Cyclase 1 (AC1) activation in synaptosomes and exerts a neuroprotective effect in vivo. It also possesses anti-inflammatory activity.
  • Suppresses glutamate release.
  • Decreases Ca2+/calmodulin/adenylate Cyclase 1 (AC1) activation.
  • Provides a neuroprotective effect in vivo.
  • Exhibits anti-inflammatory activity.
Catalog No. 50-004-16580 Supplier Medchemexpress LLC Supplier No. HYN107721MG
Albanin A, a flavonoid, suppresses glutamate release by decreasing Ca2+/calmodulin/adenylate Cyclase 1 (AC1) activation in synaptosomes and exerts a neuroprotective effect in vivo. It also possesses anti-inflammatory activity.
  • Suppresses glutamate release.
  • Decreases Ca2+/calmodulin/adenylate Cyclase 1 (AC1) activation.
  • Provides a neuroprotective effect in vivo.
  • Exhibits anti-inflammatory activity.