Learn More
Medchemexpress LLC 4H-1-Benzopyran-4-one, 2-(2-phenylethyl)- | 61828-53-3 | 25 MG
SDP
Catalog No. 5000420412
Encompass_Preferred
2-(2-Phenylethyl) chromone (Flidersiachromone) is a chromone derivative and an antioxidant. It scavenges ABTS·+ free radicals, and its derivatives show various activities such as anti-inflammatory, neuroprotective, anti-tumor, and α-glucosidase inhibitory effects. This product is suitable for experimental research in areas like inflammation-related diseases, tumors, and neurodegenerative diseases.
  • Chromone derivative and antioxidant
  • Scavenges ABTS·+ free radicals
  • Exhibits anti-inflammatory activity
  • Exhibits neuroprotective activity
  • Exhibits anti-tumor activity
  • Exhibits α-glucosidase inhibitory effects
  • Suitable for research in inflammation-related diseases
  • Suitable for research in tumors
  • Suitable for research in neurodegenerative diseases
Catalog No. 50-004-20412 Supplier Medchemexpress LLC Supplier No. 25MGHYN8220
2-(2-Phenylethyl) chromone (Flidersiachromone) is a chromone derivative and an antioxidant. It scavenges ABTS·+ free radicals, and its derivatives show various activities such as anti-inflammatory, neuroprotective, anti-tumor, and α-glucosidase inhibitory effects. This product is suitable for experimental research in areas like inflammation-related diseases, tumors, and neurodegenerative diseases.
  • Chromone derivative and antioxidant
  • Scavenges ABTS·+ free radicals
  • Exhibits anti-inflammatory activity
  • Exhibits neuroprotective activity
  • Exhibits anti-tumor activity
  • Exhibits α-glucosidase inhibitory effects
  • Suitable for research in inflammation-related diseases
  • Suitable for research in tumors
  • Suitable for research in neurodegenerative diseases