Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4H-1-benzopyran-4-one, 2,3-dihydro-5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-, (2S)- | 520-33-2 | MFCD00075646 | 99.12% | 302.28 | 100 MG
SDP

Catalog No. 5000427402
Encompass_Preferred

Hesperetin is a natural flavanone found in citrus that acts as a potent, orally active, broad-spectrum inhibitor against human UGT activity. It induces apoptosis through p38 MAPK activation and displays various bioactivities, including antioxidant, anti-inflammatory, and anti-cancer properties. Hesperetin can induce cell-cycle arrest at the G2/M phase, reduce Bcl-2, enhance BaxM, and inhibit the NF-κB receptor to induce apoptosis.

  • Purity: 99.12%
  • CAS no.: 520-33-2
  • Appearance: solid
  • Color: white to off-white
  • Molecular weight: 302.28
  • Formula: C16H14O6
  • Structure classification: flavonoids, flavanones
  • Initial source: plants other families
  • Storage: powder at -20°C for 3 years, 4°C for 2 years. In solvent at -80°C for 6 months, -20°C for 1 month.
  • Solubility: in DMSO, 125 mg/mL (413.52 mM).

Catalog No. 50-004-27402 Supplier Medchemexpress LLC Supplier No. HYN0168100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hesperetin is a natural flavanone found in citrus that acts as a potent, orally active, broad-spectrum inhibitor against human UGT activity. It induces apoptosis through p38 MAPK activation and displays various bioactivities, including antioxidant, anti-inflammatory, and anti-cancer properties. Hesperetin can induce cell-cycle arrest at the G2/M phase, reduce Bcl-2, enhance BaxM, and inhibit the NF-κB receptor to induce apoptosis.

  • Purity: 99.12%
  • CAS no.: 520-33-2
  • Appearance: solid
  • Color: white to off-white
  • Molecular weight: 302.28
  • Formula: C16H14O6
  • Structure classification: flavonoids, flavanones
  • Initial source: plants other families
  • Storage: powder at -20°C for 3 years, 4°C for 2 years. In solvent at -80°C for 6 months, -20°C for 1 month.
  • Solubility: in DMSO, 125 mg/mL (413.52 mM).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.