Learn More
Medchemexpress LLC 4H-1-benzopyran-4-one, 2-(4-hydroxyphenyl)- | 4143-63-9 | MFCD00017687 | 97.0% | 238.24 | C15H10O3 | 50 MG
SDP
Catalog No. 5000280659
Encompass_Preferred
4′-Hydroxyflavone is a flavone derivative supplied as a research-grade chemical for biochemical and pharmacological studies. It inhibits the enzyme aldose reductase (reported IC50 12 μM) and is provided with batch-specific documentation for laboratory use only.
  • Inhibits aldose reductase (IC50 = 12 μM).
  • High purity: 97.0%.
  • Molecular formula C15H10O3; molecular weight 238.24.
  • Supplied as a small-molecule reagent for in vitro studies.
  • Datasheet and certificate of analysis available.
Catalog No. 50-002-80659 Supplier Medchemexpress LLC Supplier No. HYW31909450MG
4′-Hydroxyflavone is a flavone derivative supplied as a research-grade chemical for biochemical and pharmacological studies. It inhibits the enzyme aldose reductase (reported IC50 12 μM) and is provided with batch-specific documentation for laboratory use only.
  • Inhibits aldose reductase (IC50 = 12 μM).
  • High purity: 97.0%.
  • Molecular formula C15H10O3; molecular weight 238.24.
  • Supplied as a small-molecule reagent for in vitro studies.
  • Datasheet and certificate of analysis available.