Learn More
Medchemexpress LLC 4H-1-benzopyran-4-one, 2-(4-hydroxyphenyl)- | 4143-63-9 | MFCD00017687 | 97.0% | 238.24 g/mol | C15H10O3 | 25 MG
SDP
Catalog No. 5000282513
Encompass_Preferred
4′-Hydroxyflavone is a flavone-class small molecule for biochemical and pharmacological research. It is a reported aldose reductase inhibitor (IC50 = 12 μM) and is supplied as a purified solid suitable for in vitro assays, target validation, and screening studies. Typical analytical data include CAS 4143-63-9, molecular formula C15H10O3, and molecular weight 238.24 g/mol.
  • Reported aldose reductase inhibitor (IC50 = 12 μM).
  • High purity solid (97.0%).
  • Characterized by CAS 4143-63-9 and molecular formula C15H10O3.
  • Suitable for biochemical assays, target validation, and screening.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-82513 Supplier Medchemexpress LLC Supplier No. HYW31909425MG
4′-Hydroxyflavone is a flavone-class small molecule for biochemical and pharmacological research. It is a reported aldose reductase inhibitor (IC50 = 12 μM) and is supplied as a purified solid suitable for in vitro assays, target validation, and screening studies. Typical analytical data include CAS 4143-63-9, molecular formula C15H10O3, and molecular weight 238.24 g/mol.
  • Reported aldose reductase inhibitor (IC50 = 12 μM).
  • High purity solid (97.0%).
  • Characterized by CAS 4143-63-9 and molecular formula C15H10O3.
  • Suitable for biochemical assays, target validation, and screening.
  • Available in small research pack sizes for laboratory use.