Learn More
Medchemexpress LLC 4h-1-Benzopyran-4-one, 3-(4-hydroxyphenyl)-7-methoxy-d3 | 10 MG
SDP
Catalog No. 5000419935
Encompass_Preferred
Isoformononetin-d3 is the deuterium labeled Isoformononetin. It is an analog of Daidzein and has immunoprotective effects. Isoformononetin inhibits the differentiation of Th17 and B-cells lymphopoiesis to promote osteogenesis in estrogen-deficient bone loss conditions. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules as tracers for quantitation during drug development. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Analog of daidzein
  • Immunoprotective effects
  • Inhibits differentiation of Th17 and B-cells lymphopoiesis
  • Promotes osteogenesis in estrogen-deficient bone loss conditions
  • Used as a tracer for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-19935 Supplier Medchemexpress LLC Supplier No. HYN7501S10MG
Isoformononetin-d3 is the deuterium labeled Isoformononetin. It is an analog of Daidzein and has immunoprotective effects. Isoformononetin inhibits the differentiation of Th17 and B-cells lymphopoiesis to promote osteogenesis in estrogen-deficient bone loss conditions. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules as tracers for quantitation during drug development. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Analog of daidzein
  • Immunoprotective effects
  • Inhibits differentiation of Th17 and B-cells lymphopoiesis
  • Promotes osteogenesis in estrogen-deficient bone loss conditions
  • Used as a tracer for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs