Learn More
Medchemexpress LLC 4H-1-benzopyran-4-one, 5,7-dihydroxy-2-(4-hydroxyphenyl)- | 520-36-5 | 270.24 | 200 MG
SDP
Catalog No. 5000415029
Encompass_Preferred
Apigenin (4',5,7-Trihydroxyflavone) is a competitive CYP2C9 inhibitor with a Ki of 2 μM. It inhibits cytochrome P450 2C9 (CYP2C9) with a Ki of 2 μM in the CYP2C9 RECO system, a purified, reconstituted enzyme system. This natural flavonoid possesses broad biological properties including antioxidative, anti-inflammatory, anticancer, and neuroprotective effects.
  • Inhibits cell proliferation
  • Suppresses clone formation of SGC-7901 cells
  • Alleviates Adriamycin-induced myocardial injury
  • Reduces serum aspartate amino transferase (AST) release
  • Reduces serum lactate dehydrogenase (LDH) release
  • Reduces serum creatine kinase (CK) contents
Catalog No. 50-004-15029 Supplier Medchemexpress LLC Supplier No. HYN1201200MG
Apigenin (4',5,7-Trihydroxyflavone) is a competitive CYP2C9 inhibitor with a Ki of 2 μM. It inhibits cytochrome P450 2C9 (CYP2C9) with a Ki of 2 μM in the CYP2C9 RECO system, a purified, reconstituted enzyme system. This natural flavonoid possesses broad biological properties including antioxidative, anti-inflammatory, anticancer, and neuroprotective effects.
  • Inhibits cell proliferation
  • Suppresses clone formation of SGC-7901 cells
  • Alleviates Adriamycin-induced myocardial injury
  • Reduces serum aspartate amino transferase (AST) release
  • Reduces serum lactate dehydrogenase (LDH) release
  • Reduces serum creatine kinase (CK) contents