Learn More
Medchemexpress LLC 4H-1-Benzopyran-4-one, 7-[[6-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-5-hydroxy-2-(4-methoxyphenyl)- | 480-36-4 | 592.55 | 20 MG
SDP
Catalog No. 5000388766
Encompass_Preferred
Linarin (Buddleoside) is an orally active and selective inhibitor of acetylcholinesterase (AChE). It exhibits various activities including anti-inflammatory, antioxidant, sleep aid and sedation, bone differentiation, anti-tumor, antibacterial, and antiviral properties. Linarin can be utilized in the study of diseases affecting the nervous system, osteoporosis, and cancer.
  • Orally active and selective inhibitor of acetylcholinesterase (AChE).
  • Anti-inflammatory properties.
  • Antioxidant properties.
  • Sleep aid and sedation.
  • Promotes bone differentiation.
  • Anti-tumor activity.
  • Antibacterial properties.
  • Antiviral properties.
  • Useful for studying diseases such as those of the nervous system, osteoporosis, and cancer.
Catalog No. 50-003-88766 Supplier Medchemexpress LLC Supplier No. HYN052820MG
Linarin (Buddleoside) is an orally active and selective inhibitor of acetylcholinesterase (AChE). It exhibits various activities including anti-inflammatory, antioxidant, sleep aid and sedation, bone differentiation, anti-tumor, antibacterial, and antiviral properties. Linarin can be utilized in the study of diseases affecting the nervous system, osteoporosis, and cancer.
  • Orally active and selective inhibitor of acetylcholinesterase (AChE).
  • Anti-inflammatory properties.
  • Antioxidant properties.
  • Sleep aid and sedation.
  • Promotes bone differentiation.
  • Anti-tumor activity.
  • Antibacterial properties.
  • Antiviral properties.
  • Useful for studying diseases such as those of the nervous system, osteoporosis, and cancer.