Learn More
Medchemexpress LLC 4H-1-benzothiopyran-4-one, 2,3-dihydro-2-[1-(phenylmethyl)-4-piperidinyl]- | 1204401-49-9 | MFCD18206882 | 99.9% | 337.48 g/mol | C21H23NOS | 10 MG
SDP
Catalog No. 5000242751
Encompass_Preferred
σ1 Receptor antagonist-1 is a potent and selective sigma-1 receptor antagonist provided as a solid research chemical for in vitro studies. It has high reported affinity for the σ1 receptor and has been used in cell-based assays to inhibit cell growth, induce apoptosis, and arrest the cell cycle. Analytical data and handling information are provided by the supplier; this compound is for research use only.
Catalog No. 50-002-42751 Supplier Medchemexpress LLC Supplier No. HY1081510MG
Add to Cart
Add to Cart
σ1 Receptor antagonist-1 is a potent and selective sigma-1 receptor antagonist provided as a solid research chemical for in vitro studies. It has high reported affinity for the σ1 receptor and has been used in cell-based assays to inhibit cell growth, induce apoptosis, and arrest the cell cycle. Analytical data and handling information are provided by the supplier; this compound is for research use only.