Learn More
Medchemexpress LLC 4H,8H-Benzo[1,2-b:3,4-b']dipyran-4-one, 9,10-dihydro-3,5-dihydroxy-2-(4-methoxyphenyl)-8,8-dimethyl | 38226-86-7 | 368.38 | 5 MG
SDP
Catalog No. 5000416426
Encompass_Preferred
β-Anhydroicaritin is isolated from Boswellia carterii Birdware and possesses important biological and pharmacological effects. It ameliorates the degradation of periodontal tissue, inhibits the synthesis and secretion of TNF-α and MMP-3 in diabetic rats, and decreases the overproduction of NO, IL-10, TNF-α, MCP-1, and IL-6 in peritonitis mice. Additionally, it inhibits the elevation of intracellular Ca2+ and markedly decreases iNOS protein expression.
  • Antiosteoporosis properties
  • Estrogen regulation properties
  • Antitumor properties
  • Ameliorates the degradation of periodontal tissue
  • Inhibits the synthesis and secretion of TNF-α and MMP-3 in diabetic rats
  • Decreases the overproduction of NO, IL-10, TNF-α, MCP-1, and IL-6 in peritonitis mice
  • Inhibits the elevation of intracellular Ca2+
  • Markedly decreases iNOS protein expression
Catalog No. 50-004-16426 Supplier Medchemexpress LLC Supplier No. HYN19405MG
β-Anhydroicaritin is isolated from Boswellia carterii Birdware and possesses important biological and pharmacological effects. It ameliorates the degradation of periodontal tissue, inhibits the synthesis and secretion of TNF-α and MMP-3 in diabetic rats, and decreases the overproduction of NO, IL-10, TNF-α, MCP-1, and IL-6 in peritonitis mice. Additionally, it inhibits the elevation of intracellular Ca2+ and markedly decreases iNOS protein expression.
  • Antiosteoporosis properties
  • Estrogen regulation properties
  • Antitumor properties
  • Ameliorates the degradation of periodontal tissue
  • Inhibits the synthesis and secretion of TNF-α and MMP-3 in diabetic rats
  • Decreases the overproduction of NO, IL-10, TNF-α, MCP-1, and IL-6 in peritonitis mice
  • Inhibits the elevation of intracellular Ca2+
  • Markedly decreases iNOS protein expression