Learn More
Sigma Aldrich 4H-Benzo[d][1,3]dioxin-6-amine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- (triethylsilyl)acetylene, a bulky trialkylsilyl-protected alkyne, undergoes cadiot-chodkiewicz cross-coupling reaction with various bromoalkynes to yield synthetically useful unsymmetrical diynes. rhodium-catalyzed dimerization of (triethylsilyl)acetylene has been reported.(triethylsilyl)acetylene may be used in the synthesis of triethylsilylethynyl anthradithiophenes, such as :
- 2,8-dimethyl-5,11-bis(triethylsilylethynyl)adt (adt= anthradithiophene)
- 2,8-diethyl-5,11-bis(triethylsilylethynyl)adt
- 2,8-dipropyl-5,11-bis(triethylsilylethynyl)adt
Specifications
Specifications
| CAS | 22791-64-6 |
| Molecular Formula | C8H9NO2 |
| Linear Formula | C8H9NO2 |
| MDL Number | MFCD00047632 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 151.16 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.