Learn More
Medchemexpress LLC 4H-Cyclohepta[b]thiophene-3-carboxamide, 2-[(2-bromo-2,2-difluoroacetyl)amino]-5,6,7,8-tetrahydro-N-(2-methylphenyl)- | 2098490-06-1 | C19H19BrF2N2O2S | 5 MG
SDP
Catalog No. 5000372852
Encompass_Preferred
ANO1-IN-4 (Compound 10bm) is a reversible inhibitor for calcium-activated chloride channel transmembrane protein 16A (TMEM16A, also known as ANO1) with an IC50 of 0.030 μM. It demonstrates good metabolic stability in rat liver microsomes and inhibits spontaneous contraction in mouse isolated ileum.
  • Reversible inhibitor for calcium-activated chloride channel transmembrane protein 16A (ANO1)
  • IC50 of 0.030 μM
  • Good metabolic stability in rat liver microsomes
  • Inhibits spontaneous contraction in mouse isolated ileum
Catalog No. 50-003-72852 Supplier Medchemexpress LLC Supplier No. HY1535085MG
ANO1-IN-4 (Compound 10bm) is a reversible inhibitor for calcium-activated chloride channel transmembrane protein 16A (TMEM16A, also known as ANO1) with an IC50 of 0.030 μM. It demonstrates good metabolic stability in rat liver microsomes and inhibits spontaneous contraction in mouse isolated ileum.
  • Reversible inhibitor for calcium-activated chloride channel transmembrane protein 16A (ANO1)
  • IC50 of 0.030 μM
  • Good metabolic stability in rat liver microsomes
  • Inhibits spontaneous contraction in mouse isolated ileum