Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4H-Cyclopenta[1,2-b:5,4-b']dithiophen-4-one 98.0+%, TCI America™
SDP

Catalog No. C2715200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2715200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2715200MG Supplier TCI America Supplier No. C2715200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25796-77-4
Molecular Formula C9H4OS2
Molecular Weight (g/mol) 192.25
MDL Number MFCD16619150
InChI Key HFIUHKXJUKKOIZ-UHFFFAOYSA-N
PubChem CID 10976338
IUPAC Name 3,11-dithiatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraen-7-one
SMILES O=C1C2=C(SC=C2)C2=C1C=CS2

Specifications

Melting Point 136°C
Quantity 200 mg
Formula Weight 192.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4H-Cyclopenta[1,2-b:5,4-b′]dithiophen-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.