Learn More
Medchemexpress LLC 4H-cyclopenta[b]thiophene-2-carboxylic acid, 5,6-dihydro- | 40133-06-0 | MFCD01860021 | 99.0% | 168.21 g·mol⁻¹ | C8H8O2S | 250 MG
SDP
Catalog No. 5000208460
Encompass_Preferred
PRL3-CNNM4 PPI-IN-1 is a small-molecule inhibitor of the PRL3-CNNM4 protein-protein interaction (Compound C28d52) used as a research tool to study PRL3-mediated regulation of CNNM proteins. The compound is reported to penetrate epithelial cell layers, show metabolic stability, and possess favorable pharmacokinetic and pharmacodynamic properties.
  • Inhibits PRL3-CNNM4 protein interaction.
  • Penetrates epithelial cell layers.
  • Exhibits metabolic stability and favorable PK/PD properties.
  • High reported purity (~99%).
  • Available in research-scale pack sizes, including 250 mg.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-08460 Supplier Medchemexpress LLC Supplier No. 250MGHYW043170
PRL3-CNNM4 PPI-IN-1 is a small-molecule inhibitor of the PRL3-CNNM4 protein-protein interaction (Compound C28d52) used as a research tool to study PRL3-mediated regulation of CNNM proteins. The compound is reported to penetrate epithelial cell layers, show metabolic stability, and possess favorable pharmacokinetic and pharmacodynamic properties.
  • Inhibits PRL3-CNNM4 protein interaction.
  • Penetrates epithelial cell layers.
  • Exhibits metabolic stability and favorable PK/PD properties.
  • High reported purity (~99%).
  • Available in research-scale pack sizes, including 250 mg.
  • For research use only; not for human or clinical use.