Learn More
Medchemexpress LLC 4H-furo[3,2-b]pyrrole-5-carboxylic acid | 67268-37-5 | MFCD07368660 | 99.4% | 151.12 g/mol | C7H5NO3 | 250 MG
SDP
Catalog No. 5000302392
Encompass_Preferred
This compound is a small-molecule research chemical that functions as an orally active and potent D-amino acid oxidase (DAAO) inhibitor. It is supplied as a solid (gray to brown), with molecular formula C7H5NO3 and molecular weight 151.12 g/mol, intended for biochemical and pharmacological research applications.
  • Orally active DAAO inhibitor suitable for in vitro and in vivo studies.
  • High purity reported at 99.44% for reliable experimental results.
  • Solid form, gray to brown, convenient for storage and handling.
  • Molecular formula C7H5NO3 and molecular weight 151.12 g/mol.
  • Available in small research quantities (100 mg-1 g), larger amounts available by request.
Catalog No. 50-003-02392 Supplier Medchemexpress LLC Supplier No. 250MGHYW027382
This compound is a small-molecule research chemical that functions as an orally active and potent D-amino acid oxidase (DAAO) inhibitor. It is supplied as a solid (gray to brown), with molecular formula C7H5NO3 and molecular weight 151.12 g/mol, intended for biochemical and pharmacological research applications.
  • Orally active DAAO inhibitor suitable for in vitro and in vivo studies.
  • High purity reported at 99.44% for reliable experimental results.
  • Solid form, gray to brown, convenient for storage and handling.
  • Molecular formula C7H5NO3 and molecular weight 151.12 g/mol.
  • Available in small research quantities (100 mg-1 g), larger amounts available by request.