Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4H-imidazo[1,5-a]benzodiazepine-3-carboxylic acid, 8-fluoro-5,6-dihydro-5-methyl-6-oxo- | 84378-44-9 | 100.0% | 275.24 | C13H10FN3O3 | 1 ML
SDP

Catalog No. 5000229210
Encompass_Preferred

Flumazenil acid is the carboxylic acid metabolite of flumazenil, provided as a high-purity reference material for research on GABAA receptor pharmacology and drug metabolism. It is supplied as a powder and is commonly prepared as a 10 mM solution (1 mL) for in vitro studies.

  • High purity: 99.96% as reported by certificate of analysis.
  • Molecular formula C13H10FN3O3 and molecular weight 275.24 g/mol.
  • Available as powder; solutions commonly prepared at 10 mM, 1 mL volumes.
  • Storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).
  • Intended for research use in pharmacology and metabolism studies (in vitro).

Catalog No. 50-002-29210 Supplier Medchemexpress LLC Supplier No. HY11884410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flumazenil acid is the carboxylic acid metabolite of flumazenil, provided as a high-purity reference material for research on GABAA receptor pharmacology and drug metabolism. It is supplied as a powder and is commonly prepared as a 10 mM solution (1 mL) for in vitro studies.

  • High purity: 99.96% as reported by certificate of analysis.
  • Molecular formula C13H10FN3O3 and molecular weight 275.24 g/mol.
  • Available as powder; solutions commonly prepared at 10 mM, 1 mL volumes.
  • Storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).
  • Intended for research use in pharmacology and metabolism studies (in vitro).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.