Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4H-indol-4-one, 1-[2-[4-(3-chlorophenyl)-1-piperazinyl]ethyl]-1,5,6,7-tetrahydro- | 496921-73-4 | 99.2% | 357.88 g/mol | C20H24ClN3O | 1 ML
SDP

Catalog No. 5000248305
Encompass_Preferred

NEO 376 is a research-grade small molecule reported as a selective modulator of 5-HT1, GABA, and dopamine receptors with documented antipsychotic activity. It is offered as a solid and as a ready-to-use 10 mM solution in DMSO for in vitro research; handle and store according to supplied safety and handling documents.

  • Selective modulator of 5-HT1, GABA, and dopamine receptors.
  • Provided as 10 mM solution in DMSO and as solid formats (typical milligram packs).
  • High purity (~99.2%).
  • Molecular weight 357.88 g/mol; formula C20H24ClN3O.
  • Storage: powder -20°C (long term); in solvent -80°C for extended storage.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-48305 Supplier Medchemexpress LLC Supplier No. HY10158310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NEO 376 is a research-grade small molecule reported as a selective modulator of 5-HT1, GABA, and dopamine receptors with documented antipsychotic activity. It is offered as a solid and as a ready-to-use 10 mM solution in DMSO for in vitro research; handle and store according to supplied safety and handling documents.

  • Selective modulator of 5-HT1, GABA, and dopamine receptors.
  • Provided as 10 mM solution in DMSO and as solid formats (typical milligram packs).
  • High purity (~99.2%).
  • Molecular weight 357.88 g/mol; formula C20H24ClN3O.
  • Storage: powder -20°C (long term); in solvent -80°C for extended storage.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.