Learn More
Medchemexpress LLC 4H-Indol-4-one, 3-ethyl-1,5,6,7-tetrahydro-2-methyl-5-(4-morpholinylmethyl)-, hydrochloride (1:1) | 15622-65-8 | 99.8% | 1 ML
SDP
Catalog No. 5000407464
Encompass_Preferred
Molindone hydrochloride is an orally active and brain-penetrant dopamine D2/D5 receptor antagonist. It exhibits antipsychotic and antidepressant-like activities, suppressing spontaneous locomotion and antagonizing apomorphine-induced emesis. It can be used for research related to neurological diseases.
  • Orally active and brain-penetrant dopamine D2/D5 receptor antagonist.
  • Shows antipsychotic and antidepressant-like activities.
  • Suppresses spontaneous locomotion.
  • Antagonizes apomorphine-induced emesis.
  • Can be used for the research of neurological disease.
Catalog No. 50-004-07464 Supplier Medchemexpress LLC Supplier No. HYB101710MM/1ML
Molindone hydrochloride is an orally active and brain-penetrant dopamine D2/D5 receptor antagonist. It exhibits antipsychotic and antidepressant-like activities, suppressing spontaneous locomotion and antagonizing apomorphine-induced emesis. It can be used for research related to neurological diseases.
  • Orally active and brain-penetrant dopamine D2/D5 receptor antagonist.
  • Shows antipsychotic and antidepressant-like activities.
  • Suppresses spontaneous locomotion.
  • Antagonizes apomorphine-induced emesis.
  • Can be used for the research of neurological disease.