Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4H-Pyran-4-one

Catalog No. 1772291G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-722-91G 1 g
17-722-95G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-722-91G Supplier Sigma Aldrich Supplier No. 1772291G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4H-Pyran-4-one

Specifications

CAS 108-97-4
Melting Point 32°C to 34°C (lit.)
Boiling Point 210°C to 215°C (lit.)
Molecular Formula C5H4O2
Linear Formula C5H4O2
MDL Number MFCD00006576
Quantity 1 g
Synonym gamma-Pyrone
Molecular Weight (g/mol) 96.08
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.