Learn More
Medchemexpress LLC 4H-Pyrazino[2,1-a]isoquinolin-4-one, 2-(cyclohexyl-1,2,2,3,3,4,4,5,5,6,6-d11-carbonyl)-1,2,3,6,7 | 1246343-36-1 | 99.6% | 1 MG
SDP
Catalog No. 5000405597
Encompass_Preferred
Praziquantel-d11 is the deuterium labeled Praziquantel, an anthelmintic compound. It serves as a tracer and internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled anthelmintic
  • Functions as a tracer for research applications
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05597 Supplier Medchemexpress LLC Supplier No. HYB0244S1MG
Praziquantel-d11 is the deuterium labeled Praziquantel, an anthelmintic compound. It serves as a tracer and internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled anthelmintic
  • Functions as a tracer for research applications
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS